Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-tert-Butylanthracene ≥98.0%

Supplier: TCI America

B3541-5G B3541-1G
TCB3541-1GEA 110.59 USD InStock
TCB3541-1G TCB3541-5G
2-tert-Butylanthracene ≥98.0%
2-tert-Butylanthracene
CAS Number: 18801-00-8
MDL Number: MFCD00003581
Molecular Formula: C18H18
Molecular Weight: 234.34
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 148


Formula: C₁₈H₁₈
MW: 234.34 g/mol
Boiling Pt: 363 °C (1013 hPa)
Melting Pt: 148 °C
MDL Number: MFCD00003581
CAS Number: 18801-00-8
EINECS: 242-588-3
https://us-legacy.vwr.com/stibo/web/std.lang.all/48/80/28934880.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty