Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

trans-Cinnamic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning
Synonyms: trans-Cinnamic acid

C0353-500G C0353-100G C0353-25G
TCC0353-500GEA 81.5 USD InStock
TCC0353-500G TCC0353-025G TCC0353-100G
trans-Cinnamic acid ≥98.0% (by GC, titration analysis)
trans-Cinnamic acid
CAS Number: 140-10-3
MDL Number: MFCD00004369
Molecular Formula: C9H8O2
Molecular Weight: 148.16
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Boiling point (°C): 300
Melting point (°C): 135
Lambda max.: 273 nm (MeOH)

Product Categories: Synthetic Organic Chemistry, Carboxylic Acids (Multi-component Reaction), Bioscience, Crude Medicine Ingredients for Research and Experimental Use, Phenylpropanoids, Phenylpropanoids (Phytochemicals), Analytical Science, Standards for Mass Spectrometry &
Other Product Names / Synonyms: trans-3-Phenylacrylic Acid


Formula: C₉H₈O₂
MW: 148.16 g/mol
Boiling Pt: 300 °C
Melting Pt: 135 °C
Density: 1.25 g/cm³ (4 °C)
MDL Number: MFCD00004369
CAS Number: 140-10-3
EINECS: 205-398-1
https://us-legacy.vwr.com/stibo/web/std.lang.all/73/89/28937389.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC,T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty