C0449-1G
C0449-100MG
TCC0449-100MGEA
48.28
USD
InStock
TCC0449-100MG
TCC0449-001G
Vitamin B12 (cyanocobalamin) ≥95.0% (by UV method)
Vitamin B12 (cyanocobalamin)
CAS Number: 68-19-9
MDL Number: MFCD00151092
Molecular Formula: C63H88CoN14O14P
Molecular Weight: 1355.39
Purity/Analysis Method: 95.0% (E)
Form: Crystal
Storage Temperature: 0-10°C
Product Categories: Bioscience, Porphyrins, Vitamins
Other Product Names / Synonyms: Cyanocobalamin