Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl-2-chloroacetoacetate ≥95.0%

Supplier: TCI America
Synonyms: Ethyl 2-chloroacetoacetate

C1218-25G C1218-500G
TCC1218-25GEA 21.16 USD InStock
TCC1218-25G TCC1218-500G
Ethyl-2-chloroacetoacetate ≥95.0%
Ethyl 2-chloroacetoacetate
CAS Number: 609-15-4
MDL Number: MFCD00009141
Molecular Formula: C6H9ClO3
Molecular Weight: 164.59
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 77
Flash Point (°C): 54
Specific Gravity (20/20): 1.18

Other Product Names / Synonyms: 2-Chloroacetoacetic Acid Ethyl Ester


Formula: C₆H₉ClO₃
MW: 164.59 g/mol
Boiling Pt: 77 °C
Melting Pt: –80 °C
Density: 1.18 (20 °C)
Flash Pt: 54 °C
MDL Number: MFCD00009141
CAS Number: 609-15-4
EINECS: 210-180-4
https://us-legacy.vwr.com/stibo/web/std.lang.all/84/64/28938464.jpg
Order Now
Specification Test Results
Purity/Analysis Method 95.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty