Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-(4-Chlorobutoxy)tetrahydropyran ≥95.0%

Supplier: TCI America

C1279-5G C1279-25G C1279-1G
TCC1279-005GEA 142 USD InStock
TCC1279-005G 76219-670 TCC1279-25G
2-(4-Chlorobutoxy)tetrahydropyran ≥95.0%
2-(4-Chlorobutoxy)tetrahydropyran
CAS Number: 41302-05-0
MDL Number: MFCD00142812
Molecular Formula: C9H17ClO2
Molecular Weight: 192.68
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 131
Specific Gravity (20/20): 1.07


Formula: C₉H₁₇ClO₂
Boiling Pt: 131 °C
Density: 1.07 (20 °C)
MDL Number: MFCD00142812
CAS Number: 41302-05-0
https://us-legacy.vwr.com/stibo/web/std.lang.all/85/21/28938521.jpg
Order Now
Specification Test Results
Purity/Analysis Method 95.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty