Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Carboxy-2,2,5,5-tetramethylpyrrolidine-1-oxyl ≥98.0% (by HPLC, titration analysis) free radical

Supplier: TCI America
Synonyms: 3-Carboxy-PROXYL

C1406-1G
TCC1406-001GEA 354.52 USD InStock
TCC1406-001G
3-Carboxy-2,2,5,5-tetramethylpyrrolidine-1-oxyl ≥98.0% (by HPLC, titration analysis) free radical
3-Carboxy-2,2,5,5-tetramethylpyrrolidine-1-oxyl
CAS Number: 2154-68-9
MDL Number: MFCD00003167
Molecular Formula: C9H16NO3
Molecular Weight: 186.23
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Storage Temperature: 0-10°C

Product Categories: Materials Chemistry, Organic Radicals (Magnetic Materials), Bioscience, ROS/RNS Detection Reagents, Analytical Science, Spin Labels
Other Product Names / Synonyms: 3-Carboxy-PROXYL


Formula: C₉H₁₆NO₃
MW: 186.23 g/mol
MDL Number: MFCD00003167
CAS Number: 2154-68-9
https://us-legacy.vwr.com/stibo/web/std.lang.all/86/43/28938643.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (HPLC,T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty