Synonyms:
4-Carboxy-TEMPO
C1428-1G
C1428-100MG
TCC1428-100MGEA
78.12
USD
InStock
TCC1428-100MG
TCC1428-001G
4-Carboxy-2,2,6,6-tetramethylpiperidine-1-oxyl ≥97.0% (by GC, titration analysis) free radical
4-Carboxy-2,2,6,6-tetramethylpiperidine-1-oxyl
CAS Number: 37149-18-1
MDL Number: MFCD00134569
Molecular Formula: C10H18NO3
Molecular Weight: 200.26
Purity/Analysis Method: 97.0% (GC,T)
Form: Crystal
Melting point (°C): 178
Product Categories: Materials Chemistry, Organic Radicals (Magnetic Materials), Precise Polymerization Reagents, Analytical Science, Spin Labels
Other Product Names / Synonyms: 4-Carboxy-TEMPO