Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(2-Chloroacetamido)-2,2,6,6-tetramethylpiperidine-1-oxyl ≥96.0% (by HPLC) free radical

Supplier: TCI America
Synonyms: 4-(2-Chloroacetamido)-TEMPO

C1432-1G C1432-100MG
TCC1432-100MGEA 47.69 USD InStock
TCC1432-100MG TCC1432-1G
4-(2-Chloroacetamido)-2,2,6,6-tetramethylpiperidine-1-oxyl ≥96.0% (by HPLC) free radical
4-(2-Chloroacetamido)-2,2,6,6-tetramethylpiperidine-1-oxyl
CAS Number: 36775-23-2
MDL Number: MFCD00041858
Molecular Formula: C11H20ClN2O2
Molecular Weight: 247.74
Purity/Analysis Method: 96.0% (HPLC)
Form: Crystal
Melting point (°C): 117

Product Categories: Materials Chemistry, Organic Radicals (Magnetic Materials), Analytical Science, Spin Labels
Other Product Names / Synonyms: 4-(2-Chloroacetamido)-TEMPO


Formula: C₁₁H₂₀ClN₂O₂
MW: 247.74 g/mol
Melting Pt: 117 °C
MDL Number: MFCD00041858
CAS Number: 36775-23-2
https://us-legacy.vwr.com/stibo/web/std.lang.all/86/58/28938658.jpg
Order Now
Specification Test Results
Purity/Analysis Method 96.0% (HPLC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty