Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-(Hydroxymethyl)benzonitrile ≥98.0%

Supplier: TCI America
Synonyms: 3-Cyanobenzyl alcohol

C1510-1G C1510-5G
TCC1510-001GEA 31.47 USD InStock
TCC1510-001G TCC1510-005G
3-(Hydroxymethyl)benzonitrile ≥98.0%
3-(Hydroxymethyl)benzonitrile
CAS Number: 874-97-5
MDL Number: MFCD01321087
Molecular Formula: C8H7NO
Molecular Weight: 133.15
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 165
Flash Point (°C): 109
Specific Gravity (20/20): 1.13

Other Product Names / Synonyms: 3-(Hydroxymethyl)benzonitrile


Formula: C₈H₇NO
MW: 133.15 g/mol
Boiling Pt: 165 °C
Density: 1.13 (20 °C)
Flash Pt: 109 °C
MDL Number: MFCD01321087
CAS Number: 874-97-5
https://us-legacy.vwr.com/stibo/web/std.lang.all/87/30/28938730.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty