Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4'-Chloro-2'-fluoroacetanilide ≥98.0%

Supplier: TCI America
Synonyms: N-(4-Chloro-2-fluorophenyl)acetamide

C1692-5G C1692-25G
TCC1692-25GEA 72.07 USD InStock
TCC1692-25G TCC1692-5G
4'-Chloro-2'-fluoroacetanilide ≥98.0%
4'-Chloro-2'-fluoroacetanilide
CAS Number: 59280-70-5
MDL Number: MFCD00042520
Molecular Formula: C8H7ClFNO
Molecular Weight: 187.60
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 157

Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2'-Fluoro-4'-chloroacetanilide


Formula: C₈H₇ClFNO
MW: 187.6 g/mol
Melting Pt: 157 °C
MDL Number: MFCD00042520
CAS Number: 59280-70-5
https://us-legacy.vwr.com/stibo/web/std.lang.all/89/01/28938901.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty