Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

ɑ-Cyano-4-hydroxycinnamic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Synonyms: α-CHCA2-Cyano-3-(4-hydroxyphenyl)acrylic Acidα-Cyano-4-hydroxycinnamic AcidCHCA

C1768-1G
TCC1768-1GEA 136.2 USD InStock
TCC1768-1G
ɑ-Cyano-4-hydroxycinnamic acid ≥98.0% (by HPLC, titration analysis)
alpha-Cyano-4-hydroxycinnamic acid
CAS Number: 28166-41-8
MDL Number: MFCD00004204
Molecular Formula: C10H7NO3
Molecular Weight: 189.17
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 250

Product Categories: Bioscience, Phenylpropanoids, Analytical Science, Standards for Mass Spectrometry & Matrix Materials
Other Product Names / Synonyms: 2-Cyano-3-(4-hydroxyphenyl)acrylic Acid, alpha-CHCA


Formula: C₁₀H₇NO₃
MW: 189.17 g/mol
Melting Pt: 250 °C
MDL Number: MFCD00004204
CAS Number: 28166-41-8
https://us-legacy.vwr.com/stibo/web/std.lang.all/89/58/28938958.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (HPLC,T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty