Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Cyano-2,2,6,6-tetramethylpiperidine-1-oxyl ≥95.0% (by titrimetric analysis) free radical

Supplier: TCI America
Synonyms: 4-Cyano-TEMPO

C1782-1G
TCC1782-1GEA 339.87 USD InStock
TCC1782-1G
4-Cyano-2,2,6,6-tetramethylpiperidine-1-oxyl ≥95.0% (by titrimetric analysis) free radical
4-Cyano-2,2,6,6-tetramethylpiperidine-1-oxyl
CAS Number: 38078-71-6
MDL Number: MFCD00191964
Molecular Formula: C10H17N2O
Molecular Weight: 181.26
Purity/Analysis Method: 95.0% (T)
Form: Crystal
Melting point (°C): 147
Storage Temperature: 0-10°C

Product Categories: Materials Chemistry, Organic Radicals (Magnetic Materials), Analytical Science, Spin Labels
Other Product Names / Synonyms: 4-Cyano-TEMPO


Formula: C₁₀H₁₇N₂O
MW: 181.26 g/mol
Melting Pt: 147 °C
MDL Number: MFCD00191964
CAS Number: 38078-71-6
https://us-legacy.vwr.com/stibo/web/std.lang.all/89/72/28938972.jpg
Order Now
Specification Test Results
Purity/Analysis Method 95.0% (T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty