Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chirabite-AR

Supplier: TCI America
Synonyms: (R)-2,2'-[5-Nitroisophthalamidobis(2,6-pyridylenecarbamoylmethoxy)]-1,1'-binaphthyl

C2184-100MG
TCC2184-100MGEA 285.12 USD InStock
TCC2184-100MG
Chirabite-AR
Chirabite-AR
CAS Number: 909254-56-4
Molecular Formula: C42H29N7O8
Molecular Weight: 759.74
Form: Crystal
Color: Slightly Pale Yellow
Storage Temperature: 0-10°C

Other Product Names / Synonyms: (R)-2,2'-[5-Nitroisophthalamidobis(2,6-pyridylenecarbamoylmethoxy)]-1,1'-binaphthyl


Formula: C₄₂H₂₉N₇O₈
MW: 759.74 g/mol
CAS Number: 909254-56-4
https://us-legacy.vwr.com/stibo/web/std.lang.all/92/03/28939203.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty