Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dansyl chloride

Supplier: TCI America
Synonyms: 5-(Dimethylamino)naphthalene-1-sulphonyl chloride

D0005-10ML
TCD0005-10MLEA 125.05 USD InStock
TCD0005-10ML
Dansyl chloride
Dansyl chloride
CAS Number: 605-65-2
MDL Number: MFCD00003985
Molecular Formula: C12H12ClNO2S
Molecular Weight: 269.74
Form: Clear Liquid
Color: Yellow
Storage Temperature: 0-10°C

Other Product Names / Synonyms: 5-(Dimethylamino)naphthalene-1-sulfonyl Chloride


Formula: C₁₂H₁₂ClNO₂S
MW: 269.75 g/mol
MDL Number: MFCD00003985
CAS Number: 605-65-2
UN: 3261
ADR: 8,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/43/07/28974307.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty