Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dansyl chloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Synonyms: 5-(Dimethylamino)naphthalene-1-sulphonyl chloride

D0656-1G D0656-5G D0656-25G
TCD0656-1GEA 110.16 USD InStock
TCD0656-1G TCD0656-5G TCD0656-25G
Dansyl chloride ≥98.0% (by HPLC, titration analysis)
Dansyl chloride
CAS Number: 605-65-2
MDL Number: MFCD00003985
Molecular Formula: C12H12ClNO2S
Molecular Weight: 269.74
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 70
Storage Temperature: 0-10°C

Product Categories: Bioscience, Fluorescent Labeling Dyes, Analytical Science, Derivatization Reagents for MS, Fluorescence Detection (HPLC Labeling Reagents), Fluorescence Detection (HPLC Labeling Reagents)
Other Product Names / Synonyms: 5-(Dimethylamino)naphthalene-1-sulfonyl Chloride


Formula: C₁₂H₁₂ClNO₂S
MW: 269.75 g/mol
Melting Pt: 70 °C
MDL Number: MFCD00003985
CAS Number: 605-65-2
UN: 3261
ADR: 8,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/55/37/28975537.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (HPLC,T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty