Synonyms:
2,5-Diphenyl-1,3-oxazole
, PPO
D0902-500G
D0902-100G
D0902-25G
TCD0902-500GEA
1725
USD
InStock
TCD0902-500G
TCD0902-025G
TCD0902-100G
2,5-Diphenyloxazole ≥99.0% (by HPLC, total nitrogen)
2,5-Diphenyloxazole
[for scintillation spectrometry]
CAS Number: 92-71-7
MDL Number: MFCD00005306
Molecular Formula: C15H11NO
Molecular Weight: 221.26
Purity/Analysis Method: 99.0% (HPLC,N)
Form: Crystal
Melting point (°C): 72
Product Categories: Analytical Science, Scintillators
Other Product Names / Synonyms: DPO, PPO