Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,5-Diphenyloxazole ≥99.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning
Synonyms: 2,5-Diphenyl-1,3-oxazolePPO

D0902-500G D0902-100G D0902-25G
TCD0902-500GEA 1725 USD InStock
TCD0902-500G TCD0902-025G TCD0902-100G
2,5-Diphenyloxazole ≥99.0% (by HPLC, total nitrogen)
2,5-Diphenyloxazole
[for scintillation spectrometry]
CAS Number: 92-71-7
MDL Number: MFCD00005306
Molecular Formula: C15H11NO
Molecular Weight: 221.26
Purity/Analysis Method: 99.0% (HPLC,N)
Form: Crystal
Melting point (°C): 72

Product Categories: Analytical Science, Scintillators
Other Product Names / Synonyms: DPO, PPO


Formula: C₁₅H₁₁NO
MW: 221.26 g/mol
Boiling Pt: 360 °C (1013 hPa)
Melting Pt: 72 °C
Density: 1.094 g/cm³ (99 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005306
CAS Number: 92-71-7
EINECS: 202-181-3
https://us-legacy.vwr.com/stibo/web/std.lang.all/60/30/28976030.jpg
Order Now
Specification Test Results
Purity/Analysis Method 99.0% (HPLC,N)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty