Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,3-Dibromotetrafluorobenzene ≥98.0%

Supplier: TCI America
Warning

D1492-5G D1492-1G
TCD1492-1GEA 49.66 USD InStock
TCD1492-1G TCD1492-005G
1,3-Dibromotetrafluorobenzene ≥98.0%
1,3-Dibromotetrafluorobenzene
CAS Number: 1559-87-1
MDL Number: MFCD00031533
Molecular Formula: C6Br2F4
Molecular Weight: 307.87
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Flash Point (°C): 92
Specific Gravity (20/20): 2.24


Formula: C₆Br₂F₄
MW: 307.87 g/mol
Boiling Pt: 85 °C (18 mmHg)
Density: 2.24 (20 °C)
Flash Pt: 92 °C
MDL Number: MFCD00031533
CAS Number: 1559-87-1
EINECS: 216-326-3
https://us-legacy.vwr.com/stibo/web/std.lang.all/83/14/28978314.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty