Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,3-Dimethyl-5-pyrazolone ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Synonyms: 2,4-Dihydro-2,5-dimethyl-3H-pyrazol-3-one1,3-Dimethyl-1H-pyrazol-5(4H)-one

D2031-100G D2031-500G D2031-25G
TCD2031-500GEA 429.16 USD InStock
TCD2031-500G TCD2031-100G TCD2031-25G
1,3-Dimethyl-5-pyrazolone ≥98.0% (by HPLC, titration analysis)
1,3-Dimethyl-5-pyrazolone
CAS Number: 2749-59-9
MDL Number: MFCD00067008
Molecular Formula: C5H8N2O
Molecular Weight: 112.13
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 117

Other Product Names / Synonyms: 2,4-Dihydro-2,5-dimethyl-3H-pyrazol-3-one


Formula: C₅H₈N₂O
MW: 112.13 g/mol
Melting Pt: 117 °C
MDL Number: MFCD00067008
CAS Number: 2749-59-9
https://us-legacy.vwr.com/stibo/web/std.lang.all/88/21/28978821.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (HPLC,T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty