Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl-1,3-dithiolane-2-carboxylate ≥97.0%

Supplier: TCI America
Synonyms: 1,3-Dithiolane-2-carboxylic Acid Ethyl Ester

D2105-5G
TCD2105-005GEA 67.97 USD InStock
TCD2105-005G
Ethyl-1,3-dithiolane-2-carboxylate ≥97.0%
Ethyl-1,3-dithiolane-2-carboxylate
CAS Number: 20461-99-8
MDL Number: MFCD00005411
Molecular Formula: C6H10O2S2
Molecular Weight: 178.26
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 85
Flash Point (°C): 110
Specific Gravity (20/20): 1.25

Product Categories: Synthetic Organic Chemistry, Sulfides
Other Product Names / Synonyms: 1,3-Dithiolane-2-carboxylic Acid Ethyl Ester


Formula: C₆H₁₀O₂S₂
MW: 178.26 g/mol
Boiling Pt: 85 °C
Density: 1.25 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00005411
CAS Number: 20461-99-8
https://us-legacy.vwr.com/stibo/web/std.lang.all/88/87/28978887.jpg
Order Now
Specification Test Results
Purity/Analysis Method 97.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty