Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5,5-Dimethyl-1-pyrroline-N-oxide ≥97.0% (by GC, titration analysis)

Supplier: TCI America
Synonyms: DMPO

D2362-1G D2362-5G
TCD2362-001GEA 129.86 USD InStock
TCD2362-001G TCD2362-5G
5,5-Dimethyl-1-pyrroline-N-oxide ≥97.0% (by GC, titration analysis)
5,5-Dimethyl-1-pyrroline-N-oxide
CAS Number: 3317-61-1
MDL Number: MFCD00005279
Molecular Formula: C6H11NO
Molecular Weight: 113.16
Purity/Analysis Method: 97.0% (GC,T)
Form: Crystal
Flash Point (°C): 95
Freezing point (°C): 30
Storage Temperature: 0°C

Product Categories: Bioscience, ROS/RNS Detection Reagents, Reagents for Nitric Oxide Research, Analytical Science, Spin Trapping Reagents
Other Product Names / Synonyms: DMPO


Formula: C₆H₁₁NO
MW: 113.16 g/mol
Flash Pt: 95 °C
MDL Number: MFCD00005279
CAS Number: 3317-61-1
https://us-legacy.vwr.com/stibo/web/std.lang.all/92/92/28979292.jpg
Order Now
Specification Test Results
Purity/Analysis Method 97.0% (GC,T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty