Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(Diethylamino)benzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Synonyms: 4-Diethylaminobenzoic Acid

D2493-5G D2493-25G
TCD2493-025GEA 113.23 USD InStock
TCD2493-025G TCD2493-5G
4-(Diethylamino)benzoic acid ≥98.0% (by GC, titration analysis)
4-(Diethylamino)benzoic acid
CAS Number: 5429-28-7
MDL Number: MFCD00002538
Molecular Formula: C11H15NO2
Molecular Weight: 193.25
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 196


Formula: C₁₁H₁₅NO₂
MW: 193.25 g/mol
Melting Pt: 196 °C
MDL Number: MFCD00002538
CAS Number: 5429-28-7
https://us-legacy.vwr.com/stibo/web/std.lang.all/94/19/28979419.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC,T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty