Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dichloro(1,5-cyclooctadiene)palladium(II) ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Synonyms: 1,5-Cyclooctadienepalladium(II) Dichloride

D2604-1G D2604-5G
TCD2604-001GEA 384 USD InStock
TCD2604-001G TCD2604-5G
Dichloro(1,5-cyclooctadiene)palladium(II) ≥98.0% (by titrimetric analysis)
Dichloro(1,5-cyclooctadiene)palladium(II)
CAS Number: 12107-56-1
MDL Number: MFCD00012412
Molecular Formula: C8H12Cl2Pd
Molecular Weight: 285.50
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 210

Product Categories: Synthetic Organic Chemistry, Homogeneous Catalysts, Cross-coupling Reaction using Transition Metal Catalysts
Other Product Names / Synonyms: 1,5-Cyclooctadienepalladium(II) Dichloride


Formula: C₈H₁₂Cl₂Pd
MW: 285.50 g/mol
Melting Pt: 210 °C
MDL Number: MFCD00012412
CAS Number: 12107-56-1
https://us-legacy.vwr.com/stibo/web/std.lang.all/95/74/28979574.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty