Synonyms:
2,5-DHBA
, 2,5-Dihydroxybenzoic Acid
, Gentisic acid
, Hydroquinonecarboxylic acid
, DHB
D2933-1G
D2933-5G
76219-014EA
37.94
USD
InStock
76219-014
TCD2933-5G
2,5-Dihydroxybenzoic acid ≥99.0% (by GC, titration analysis)
2,5-Dihydroxybenzoic acid
[Matrix for MALDI-TOF/MS]
CAS Number: 490-79-9
MDL Number: MFCD00002460
Molecular Formula: C7H6O4
Molecular Weight: 154.12
Purity/Analysis Method: 99.0% (GC,T)
Form: Crystal
Melting point (°C): 206
Product Categories: Analytical Science, Standards for Mass Spectrometry & Matrix Materials
Other Product Names / Synonyms: Gentisic Acid