Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,4-Difluorophenyl isothiocyanate ≥98.0%

Supplier: TCI America
Synonyms: 1,2-Difluoro-4-isothiocyanatobenzene

D4458-5G D4458-1G
TCD4458-5GEA 294.23 USD InStock
TCD4458-5G TCD4458-1G
3,4-Difluorophenyl isothiocyanate ≥98.0%
3,4-Difluorophenyl isothiocyanate
CAS Number: 113028-75-4
MDL Number: MFCD00175109
Molecular Formula: C7H3F2NS
Molecular Weight: 171.16
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 170
Specific Gravity (20/20): 1.35

Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: Isothiocyanic Acid 3,4-Difluorophenyl Ester


Formula: C₇H₃F₂NS
MW: 171.17 g/mol
Boiling Pt: 170 °C
Density: 1.35 (20 °C)
MDL Number: MFCD00175109
CAS Number: 113028-75-4
https://us-legacy.vwr.com/stibo/web/std.lang.all/65/82/28986582.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty