Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Esculetin ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Synonyms: 6,7-Dihydroxycoumarin

E0386-1G
TCE0386-001GEA 47.71 USD InStock
TCE0386-001G
Esculetin ≥98.0% (by HPLC, titration analysis)
Esculetin
CAS Number: 305-01-1
MDL Number: MFCD00006874
Molecular Formula: C9H6O4
Molecular Weight: 178.14
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal

Product Categories: Bioscience, Phenylpropanoids, Phenylpropanoids (Phytochemicals), Analytical Science, Standards for Mass Spectrometry & Matrix Materials
Other Product Names / Synonyms: 6,7-Dihydroxycoumarin


Formula: C₉H₆O₄
MW: 178.14 g/mol
MDL Number: MFCD00006874
CAS Number: 305-01-1
https://us-legacy.vwr.com/stibo/web/std.lang.all/79/09/28967909.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (HPLC,T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty