Synonyms:
2-Fluoro-1-methylpyridin-1-ium 4-methylbenzenesulphonate
, 2-Fluoro-1-methylpyridin-1-ium 4-methylbenzenesulfonate
F0225-25G
F0225-5G
TCF0225-025GEA
293.23
USD
InStock
TCF0225-025G
TCF0225-5G
2-Fluoro-1-methylpyridinium p-toluenesulphonate ≥98.0% (by titrimetric analysis)
2-Fluoro-1-methylpyridinium p-toluenesulphonate
[Fluorinating Reagent]
CAS Number: 58086-67-2
MDL Number: MFCD00011983
Molecular Formula: C13H14FNO3S
Molecular Weight: 283.32
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 132