Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Fluoro-1-methylpyridinium p-toluenesulphonate ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Synonyms: 2-Fluoro-1-methylpyridin-1-ium 4-methylbenzenesulphonate2-Fluoro-1-methylpyridin-1-ium 4-methylbenzenesulfonate

F0225-25G F0225-5G
TCF0225-025GEA 293.23 USD InStock
TCF0225-025G TCF0225-5G
2-Fluoro-1-methylpyridinium p-toluenesulphonate ≥98.0% (by titrimetric analysis)
2-Fluoro-1-methylpyridinium p-toluenesulphonate
[Fluorinating Reagent]
CAS Number: 58086-67-2
MDL Number: MFCD00011983
Molecular Formula: C13H14FNO3S
Molecular Weight: 283.32
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 132


Formula: C₁₃H₁₄FNO₃S
MW: 283.32 g/mol
Melting Pt: 132 °C
MDL Number: MFCD00011983
CAS Number: 58086-67-2
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty