Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-(5-Fluoro-2,4-dinitrophenyl)-4-methylpiperazine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Synonyms: PPZ

F0650-100MG F0650-1G
TCF0650-100MGEA 148.14 USD InStock
TCF0650-100MG TCF0650-1G
1-(5-Fluoro-2,4-dinitrophenyl)-4-methylpiperazine ≥98.0% (by HPLC, titration analysis)
1-(5-Fluoro-2,4-dinitrophenyl)-4-methylpiperazine
CAS Number: 928830-73-3
Molecular Formula: C11H13FN4O4
Molecular Weight: 284.25
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 115
Storage Temperature: 0-10°C

Product Categories: Analytical Science, Derivatization Reagents for MS
Other Product Names / Synonyms: PPZ


Formula: C₁₁H₁₃FN₄O₄
MW: 284.25 g/mol
Melting Pt: 115 °C
CAS Number: 928830-73-3
https://us-legacy.vwr.com/stibo/web/std.lang.all/91/98/28969198.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (HPLC,T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty