Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hexafluorobenzene ≥99.0%

Supplier: TCI America
Synonyms: Perfluorobenzene

H0085-5G H0085-25G H0085-250G
TCH0085-5GEA 33 USD InStock
TCH0085-5G TCH0085-25G TCH0085-250G
Hexafluorobenzene ≥99.0%
Hexafluorobenzene
CAS Number: 392-56-3
MDL Number: MFCD00000288
Molecular Formula: C6F6
Molecular Weight: 186.06
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 81
Melting point (°C): 5
Flash Point (°C): 10
Specific Gravity (20/20): 1.62

Product Categories: Synthetic Organic Chemistry, Fluorous Chemistry, Fluorinated Building Blocks, Analytical Science, Chemical Shift Standards for Multinuclear NMR
Other Product Names / Synonyms: Perfluorobenzene


Formula: C₆F₆
MW: 186.06 g/mol
Boiling Pt: 81 °C
Melting Pt: 5 °C
Density: 1.62 (20 °C)
Flash Pt: 10 °C
Storage Temperature: Ambient
MDL Number: MFCD00000288
CAS Number: 392-56-3
EINECS: 206-876-2
UN: 1993
ADR: 3,II
Merck Index: 13,04705
https://us-legacy.vwr.com/stibo/web/std.lang.all/01/48/28970148.jpg
Order Now
Specification Test Results
Purity/Analysis Method 99.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty