Synonyms:
Perfluorobenzene
H0085-5G
H0085-25G
H0085-250G
TCH0085-5GEA
33
USD
InStock
TCH0085-5G
TCH0085-25G
TCH0085-250G
Hexafluorobenzene ≥99.0%
Hexafluorobenzene
CAS Number: 392-56-3
MDL Number: MFCD00000288
Molecular Formula: C6F6
Molecular Weight: 186.06
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 81
Melting point (°C): 5
Flash Point (°C): 10
Specific Gravity (20/20): 1.62
Product Categories: Synthetic Organic Chemistry, Fluorous Chemistry, Fluorinated Building Blocks, Analytical Science, Chemical Shift Standards for Multinuclear NMR
Other Product Names / Synonyms: Perfluorobenzene