H0088-25ML
H0088-500ML
TCH0088-500MLEA
43.51
USD
InStock
TCH0088-500ML
TCH0088-25ML
Heptane (mixture of isomers) ≥99.0%
Heptane (mixture of isomers)
CAS Number: 142-82-5
MDL Number: MFCD00009544
Molecular Formula: C7H16
Molecular Weight: 100.21
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 98
Melting point (°C): -91
Flash Point (°C): -2
Specific Gravity (20/20): 0.68