Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hexamethyldisiloxane, 0.65 cSt (25°C) ≥98.0%

Supplier: TCI America
Danger
Synonyms: HMDSO Dow Corning 200® fluid/0.65 cSt Silicone fluid DC 200/0.65 cSt

H0091-100ML H0091-25ML H0091-500ML
TCH0091-500MLEA 59.06 USD InStock
TCH0091-500ML TCH0091-25ML TCH0091-100ML
Hexamethyldisiloxane, 0.65 cSt (25°C) ≥98.0%
Hexamethyldisiloxane
CAS Number: 107-46-0
MDL Number: MFCD00008265
Molecular Formula: C6H18OSi2
Molecular Weight: 162.38
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 100
Flash Point (°C): -2
Specific Gravity (20/20): 0.77


Formula: C₆H₁₈OSi₂
MW: 162.38 g/mol
Boiling Pt: 100 °C
Melting Pt: –59 °C
Density: 0.77 (20 °C)
Flash Pt: –2 °C
Storage Temperature: Ambient
MDL Number: MFCD00008265
CAS Number: 107-46-0
EINECS: 203-492-7
UN: 1993
ADR: 3,II
REACH: 01-2119496108-31
https://us-legacy.vwr.com/stibo/web/std.lang.all/01/54/28970154.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty