Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4'-Hydroxyacetophenone ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

H0193-25G H0193-500G H0193-100G
TCH0193-500GEA 216.87 USD InStock
TCH0193-500G TCH0193-100G TCH0193-025G
4'-Hydroxyacetophenone ≥98.0% (by HPLC, titration analysis)
4'-Hydroxyacetophenone
CAS Number: 99-93-4
MDL Number: MFCD00002359
Molecular Formula: C8H8O2
Molecular Weight: 136.15
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Boiling point (°C): 148
Melting point (°C): 110
Flash Point (°C): 166

Other Product Names / Synonyms: 4-Acetophenol, 4-Acetylphenol, 4-Hydroxyphenyl Methyl Ketone


Formula: C₈H₈O₂
MW: 136.15 g/mol
Boiling Pt: 148 °C
Melting Pt: 110 °C
Density: 1.139 g/cm³ (25 °C)
Flash Pt: 166 °C
Storage Temperature: Ambient
MDL Number: MFCD00002359
CAS Number: 99-93-4
EINECS: 202-802-8
https://us-legacy.vwr.com/stibo/web/std.lang.all/02/40/28970240.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (HPLC,T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty