Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.

Patton-Reeders reagent (Calconcarboxylic acid) (1:100)

Supplier: TCI America
Warning
Synonyms: 2,2'-Dihydroxy-4'-sulpho-1,1'-azonaphthalene-3-carboxylic acid Cal-Red®

H0250-25G H0250-1G
TCH0250-025GEA 82.1 USD InStock
TCH0250-025G TCH0250-001G
Patton-Reeders reagent (Calconcarboxylic acid) (1:100)
Patton-reeders reagent (Calconcarboxylic acid)
CAS Number: 3737-95-9
MDL Number: MFCD00004078
Molecular Formula: C21H14N2O7S
Molecular Weight: 438.41
Form: Crystal
Color: Deep Blue

Other Product Names / Synonyms: Calcon-carboxylic Acid, Cal Red


Formula: C₂₁H₁₄N₂O₇S
MW: 438.42 g/mol
Melting Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00004078
CAS Number: 3737-95-9
EINECS: 223-117-0
https://us-legacy.vwr.com/stibo/web/std.lang.all/02/92/28970292.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty