Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hexamethyldisilane ≥98.0%

Supplier: TCI America

H0638-10ML H0638-100ML
TCH0638-10MLEA 35.51 USD InStock
TCH0638-10ML TCH0638-100ML
Hexamethyldisilane ≥98.0%
Hexamethyldisilane
CAS Number: 1450-14-2
MDL Number: MFCD00008258
Molecular Formula: C6H18Si2
Molecular Weight: 146.38
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 113
Flash Point (°C): -9
Freezing point (°C): 13
Specific Gravity (20/20): 0.73


Formula: C₆H₁₈Si₂
Boiling Pt: 113 °C
Melting Pt: 13…15 °C
Density: 0.73 (20 °C)
Flash Pt: –9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008258
CAS Number: 1450-14-2
EINECS: 215-911-0
UN: 1993
ADR: 3,II
https://us-legacy.vwr.com/stibo/web/std.lang.all/06/49/28970649.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty