Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Hydroxy-2,2,6,6-tetramethylpiperidine 1-oxyl benzoate ≥97.0% free radical

Supplier: TCI America
Synonyms: 4-Hydroxy-TEMPO Benzoate4-Benzoyloxy-2,2,6,6-tetramethylpiperidine 1-Oxyl4-Benzoyloxy-TEMPO

H0878-1G H0878-5G
TCH0878-005GEA 244.62 USD InStock
TCH0878-005G TCH0878-001G
4-Hydroxy-2,2,6,6-tetramethylpiperidine 1-oxyl benzoate ≥97.0% free radical
4-Hydroxy-2,2,6,6-tetramethylpiperidine 1-oxyl benzoate
[Catalyst for Oxidation]
CAS Number: 3225-26-1
MDL Number: MFCD00075563
Molecular Formula: C16H22NO3
Molecular Weight: 276.36
Purity/Analysis Method: 97.0% (GC)
Form: Crystal
Melting point (°C): 104

Product Categories: Synthetic Organic Chemistry, Oxidation, Materials Chemistry, Organic Radicals (Magnetic Materials), Precise Polymerization Reagents, Analytical Science, Spin Labels
Other Product Names / Synonyms: 4-Hydroxy-TEMPO Benzoate, 4-Benzoyloxy-2,2,6,6-tetramethylpiperidine 1-Oxyl, 4-Benzoyloxy-TEMPO


Formula: C₁₆H₂₂NO₃
MW: 276.36 g/mol
Melting Pt: 104 °C
MDL Number: MFCD00075563
CAS Number: 3225-26-1
https://us-legacy.vwr.com/stibo/web/std.lang.all/09/01/28970901.jpg
Order Now
Specification Test Results
Purity/Analysis Method 97.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty