Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(2-Iodoacetamido)-2,2,6,6-tetramethylpiperidinooxy ≥98.0% (by HPLC) free radical

Supplier: TCI America
Synonyms: 4-(2-Iodoacetamido)-2,2,6,6-tetramethylpiperidine 1-Oxyl4-(2-Iodoacetamido)-TEMPO

I0487-100MG
TCI0487-100MGEA 234 USD InStock
TCI0487-100MG
4-(2-Iodoacetamido)-2,2,6,6-tetramethylpiperidinooxy ≥98.0% (by HPLC) free radical
4-(2-Iodoacetamido)-2,2,6,6-tetramethylpiperidinooxy (4-(2-Iodoacetamido)-TEMPO, free radical)
CAS Number: 25713-24-0
MDL Number: MFCD00010171
Molecular Formula: C11H20IN2O2
Molecular Weight: 339.20
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Melting point (°C): 127

Product Categories: Materials Chemistry, Organic Radicals (Magnetic Materials), Analytical Science, Spin Labels
Other Product Names / Synonyms: 4-(2-Iodoacetamido)-TEMPO


Formula: C₁₁H₂₀IN₂O₂
MW: 339.2 g/mol
Melting Pt: 127 °C
MDL Number: MFCD00010171
CAS Number: 25713-24-0
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (HPLC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty