Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Iodo-3-(trifluoromethoxy)benzene ≥98.0% (by GC)

Supplier: TCI America

I0813-5G
TCI0813-5GEA 68.01 USD InStock
TCI0813-5G
1-Iodo-3-(trifluoromethoxy)benzene ≥98.0% (by GC)
1-Iodo-3-(trifluoromethoxy)benzene
CAS Number: 198206-33-6
MDL Number: MFCD01090992
Molecular Formula: C7H4F3IO
Molecular Weight: 288.01
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 71
Flash Point (°C): 65
Specific Gravity (20/20): 1.87


Formula: C₇H₄F₃IO
MW: 288.01 g/mol
Boiling Pt: 71 °C
Density: 1,87 (20 °C)
Flash Pt: 65 °C
MDL Number: MFCD01090992
CAS Number: 198206-33-6
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty