Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Methoxy-2,2,6,6-tetramethylpiperidinooxy (4-methoxy-TEMPO) ≥98.0% (by GC, titration analysis) free radical

Supplier: TCI America
Synonyms: 4-Methoxy-2,2,6,6-tetramethyl-1-piperidinyloxy4-Methoxy-2,2,6,6-tetramethylpiperidine 1-Oxyl

M1197-5G M1197-1G
TCM1197-005GEA 154.28 USD InStock
TCM1197-005G TCM1197-1G
4-Methoxy-2,2,6,6-tetramethylpiperidinooxy (4-methoxy-TEMPO) ≥98.0% (by GC, titration analysis) free radical
4-Methoxy-2,2,6,6-tetramethylpiperidinooxy (4-Methoxy-TEMPO, free radical)
[Catalyst for Oxidation]
CAS Number: 95407-69-5
MDL Number: MFCD00270334
Molecular Formula: C10H20NO2
Molecular Weight: 186.28
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 43

Product Categories: Synthetic Organic Chemistry, Oxidation, Materials Chemistry, Organic Radicals (Magnetic Materials), Precise Polymerization Reagents, Analytical Science, Spin Labels
Other Product Names / Synonyms: 4-Methoxy-TEMPO


Formula: C₁₀H₂₀NO₂
MW: 186.27 g/mol
Melting Pt: 43 °C
MDL Number: MFCD00270334
CAS Number: 95407-69-5
https://us-legacy.vwr.com/stibo/web/std.lang.all/49/52/28974952.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC,T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty