Synonyms:
4-Methoxy-2,2,6,6-tetramethyl-1-piperidinyloxy
, 4-Methoxy-2,2,6,6-tetramethylpiperidine 1-Oxyl
M1197-5G
M1197-1G
TCM1197-005GEA
154.28
USD
InStock
TCM1197-005G
TCM1197-1G
4-Methoxy-2,2,6,6-tetramethylpiperidinooxy (4-methoxy-TEMPO) ≥98.0% (by GC, titration analysis) free radical
4-Methoxy-2,2,6,6-tetramethylpiperidinooxy (4-Methoxy-TEMPO, free radical)
[Catalyst for Oxidation]
CAS Number: 95407-69-5
MDL Number: MFCD00270334
Molecular Formula: C10H20NO2
Molecular Weight: 186.28
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 43
Product Categories: Synthetic Organic Chemistry, Oxidation, Materials Chemistry, Organic Radicals (Magnetic Materials), Precise Polymerization Reagents, Analytical Science, Spin Labels
Other Product Names / Synonyms: 4-Methoxy-TEMPO