Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Mesityl oxide ≥85.0% (contains 4-Methyl-4-penten-2-one)

Supplier: TCI America
Danger
Synonyms: 4-Methyl-3-penten-2-one

M1340-25ML M1340-500ML
TCM1340-500MLEA 63.38 USD InStock
TCM1340-500ML TCM1340-25ML
Mesityl oxide ≥85.0% (contains 4-Methyl-4-penten-2-one)
Mesityl oxide
CAS Number: 141-79-7
MDL Number: MFCD00008900
Molecular Formula: C6H10O
Molecular Weight: 98.15
Purity/Analysis Method: 85.0% (GC)
Form: Clear Liquid
Flash Point (°C): 25
Specific Gravity (20/20): 0.85

Other Product Names / Synonyms: Isopropylideneacetone, Mesityl Oxide


Formula: C₆H₁₀O
MW: 98.14 g/mol
Density: 0.85 (20 °C)
Flash Pt: 25 °C
Storage Temperature: Ambient
MDL Number: MFCD00008900
CAS Number: 141-79-7
EINECS: 205-502-5
UN: 1229
ADR: 3,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/53/12/28975312.jpg
Order Now
Specification Test Results
Purity/Analysis Method 85.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty