Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Methoxychalcone ≥98.0%

Supplier: TCI America
Synonyms: 3-(4-Methoxyphenyl)-1-phenylprop-2-en-1-one4-Methoxybenzylidenacetophenone

M1409-25G
TCM1409-25GEA 51.77 USD InStock
TCM1409-25G
4-Methoxychalcone ≥98.0%
4-Methoxychalcone
CAS Number: 959-33-1
MDL Number: MFCD00017179
Molecular Formula: C16H14O2
Molecular Weight: 238.29
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 75

Product Categories: Bioscience, Aromatic Polyketides
Other Product Names / Synonyms: 2-(p-Anisal)acetophenone, 2-(p-Anisylidene)acetophenone, 2-(4-Methoxybenzal)acetophenone, 2-(4-Methoxybenzylidene)acetophenone


Formula: C₁₆H₁₄O₂
MW: 238.29 g/mol
Melting Pt: 75 °C
MDL Number: MFCD00017179
CAS Number: 959-33-1
https://us-legacy.vwr.com/stibo/web/std.lang.all/55/46/28975546.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty