Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Methacryloyloxy-2,2,6,6-tetramethylpiperidine-1-oxyl ≥98.0% (by GC, titration analysis) radical_free

Supplier: TCI America
Synonyms: 4-Methacryloyloxy-TEMPO Free RadicalTEMPO MethacrylatePTMA Monomer

M1531-1G M1531-5G
TCM1531-1GEA 110.6 USD InStock
TCM1531-1G TCM1531-5G
4-Methacryloyloxy-2,2,6,6-tetramethylpiperidine-1-oxyl ≥98.0% (by GC, titration analysis) radical_free
4-Methacryloyloxy-2,2,6,6-tetramethylpiperidine-1-oxyl
CAS Number: 15051-46-4
MDL Number: MFCD17677357
Molecular Formula: C13H22NO3
Molecular Weight: 240.32
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 89
Storage Temperature: 0-10°C

Product Categories: Materials Chemistry, Organic Radicals (Magnetic Materials), Monomers, Analytical Science, Spin Labels
Other Product Names / Synonyms: 4-Methacryloyloxy-TEMPO Free Radical, TEMPO Methacrylate, PTMA Monomer


Formula: C₁₃H₂₂NO₃
MW: 240.32 g/mol
Melting Pt: 89 °C
MDL Number: MFCD17677357
CAS Number: 15051-46-4
https://us-legacy.vwr.com/stibo/web/std.lang.all/57/91/28975791.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC,T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty