Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Mono-6-O-(p-toluenesulfonyl)-ɑ-cyclodextrin ≥85.0% (by HPLC)

Supplier: TCI America

M1644-1G M1644-200MG
TCM1644-200MGEA 441 USD InStock
TCM1644-200MG TCM1644-1G
Mono-6-O-(p-toluenesulfonyl)-ɑ-cyclodextrin ≥85.0% (by HPLC)
Mono-6-O-(p-toluenesulfonyl)-alpha-cyclodextrin
CAS Number: 32860-56-3
Molecular Formula: C43H66O32S
Molecular Weight: 1127.03
Purity/Analysis Method: 85.0% (HPLC)
Form: Crystal
Specific rotation [a]20/D: 122 deg (C=4, DMSO)
Storage Temperature: 0°C


Formula: C₄₃H₆₆O₃₂S
MW: 1127.03 g/mol
CAS Number: 32860-56-3
https://us-legacy.vwr.com/stibo/web/std.lang.all/59/16/28975916.jpg
Order Now
Specification Test Results
Purity/Analysis Method 85.0% (HPLC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty