Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(R)-(-)-1-Methoxy-2-propanol ≥98.0% (by GC)

Supplier: TCI America
Synonyms: (R)-(-)-Monopropylene glycol methyl ether

M2166-5G M2166-25G
TCM2166-5GEA 26.39 USD InStock
TCM2166-5G TCM2166-25G
(R)-(-)-1-Methoxy-2-propanol ≥98.0% (by GC)
(R)-(-)-1-Methoxy-2-propanol
CAS Number: 4984-22-9
MDL Number: MFCD01632587
Molecular Formula: C4H10O2
Molecular Weight: 90.12
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 120
Flash Point (°C): 32
Specific Gravity (20/20): 0.92
Specific rotation [a]20/D: -22 deg (C=10, CHCl3)

Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (R)-(-)-Propylene Glycol 1-Monomethyl Ether


Formula: C₄H₁₀O₂
MW: 90.12 g/mol
Boiling Pt: 120 °C
Density: 0.92 (20 °C)
Flash Pt: 32 °C
MDL Number: MFCD01632587
CAS Number: 4984-22-9
UN: 3092
ADR: 3,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/81/42/28988142.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty