Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-(1-Naphthyl)-5-phenyloxazole ≥99.0% (by HPLC, total nitrogen)

Supplier: TCI America
Synonyms: ANPO

N0068-1G
TCN0068-001GEA 58.82 USD InStock
TCN0068-001G
2-(1-Naphthyl)-5-phenyloxazole ≥99.0% (by HPLC, total nitrogen)
2-(1-Naphthyl)-5-phenyloxazole
[for scintillation spectrometry]
CAS Number: 846-63-9
MDL Number: MFCD00019714
Molecular Formula: C19H13NO
Molecular Weight: 271.32
Purity/Analysis Method: 99.0% (HPLC,N)
Form: Crystal
Melting point (°C): 106

Product Categories: Analytical Science, Scintillators
Other Product Names / Synonyms: ANPO


Formula: C₁₉H₁₃NO
MW: 271.32 g/mol
Melting Pt: 106 °C
MDL Number: MFCD00019714
CAS Number: 846-63-9
https://us-legacy.vwr.com/stibo/web/std.lang.all/91/73/28989173.jpg
Order Now
Specification Test Results
Purity/Analysis Method 99.0% (HPLC,N)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty