Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(2-Nitrophenyl)hydrazine hydrochloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Synonyms: 2-Nitrophenylhydrazine hydrochloride

N0231-25G
TCN0231-025GEA 330.19 USD InStock
TCN0231-025G
(2-Nitrophenyl)hydrazine hydrochloride ≥98.0% (by titrimetric analysis)
(2-Nitrophenyl)hydrazine hydrochloride
CAS Number: 6293-87-4
MDL Number: MFCD00060205
Molecular Formula: C6H7N3O2
Molecular Weight: 189.60
Purity/Analysis Method: 98.0% (T)
Form: Crystal


Formula: C6H8ClN3O2
MW: 189.6 g/mol
MDL Number: MFCD00060205
CAS Number: 6293-87-4
https://us-legacy.vwr.com/stibo/web/std.lang.all/93/20/28989320.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty