Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nitrosobenzene ≥98.0%

Supplier: TCI America

N0367-25G N0367-5G
TCN0367-025GEA 225 USD InStock
TCN0367-025G TCN0367-5G
Nitrosobenzene ≥98.0%
Nitrosobenzene
CAS Number: 586-96-9
MDL Number: MFCD00002059
Molecular Formula: C6H5NO
Molecular Weight: 107.11
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 57
Melting point (°C): 66
Storage Temperature: 0-10°C


Formula: C₆H₅NO
MW: 107.11 g/mol
Boiling Pt: 57 °C
Melting Pt: 66 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002059
CAS Number: 586-96-9
EINECS: 209-591-1
UN: 2811
ADR: 6.1,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/94/50/28989450.jpg
Order Now
Specification Test Results
Purity/Analysis Method 98.0% (GC)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty