Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Oxo-2,2,6,6-tetramethylpiperidine-1-oxyl ≥95.0% (by GC, titration analysis) free radical

Supplier: TCI America
Synonyms: 4-Oxo-TEMPOTEMPONE

O0266-5G O0266-25G
TCO0266-005GEA 122.56 USD InStock
TCO0266-005G TCO0266-25G
4-Oxo-2,2,6,6-tetramethylpiperidine-1-oxyl ≥95.0% (by GC, titration analysis) free radical
4-Oxo-2,2,6,6-tetramethylpiperidine-1-oxyl
CAS Number: 2896-70-0
MDL Number: MFCD00006187
Molecular Formula: C9H16NO2
Molecular Weight: 170.23
Purity/Analysis Method: 95.0% (GC,T)
Form: Crystal
Melting point (°C): 37
Storage Temperature: 0°C

Product Categories: Materials Chemistry, Organic Radicals (Magnetic Materials), Precise Polymerization Reagents, Analytical Science, Spin Labels
Other Product Names / Synonyms: 4-Oxo-TEMPO


Formula: C₉H₁₆NO₂
MW: 170.23 g/mol
Melting Pt: 37 °C
MDL Number: MFCD00006187
CAS Number: 2896-70-0
https://us-legacy.vwr.com/stibo/web/std.lang.all/05/37/28990537.jpg
Order Now
Specification Test Results
Purity/Analysis Method 95.0% (GC,T)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty